Product Code | Brand | Quantity | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
---|---|---|---|---|---|---|---|---|
15620750
|
Honeywell Fluka
04273-1KG
|
1 kg |
£35.45
1kg |
|
||||
15630750
|
Honeywell Fluka
04273-2.5KG
|
2.5 kg |
£68.67
2.50kg |
|
||||
15650750
|
Honeywell Fluka
04273-6X1KG
|
6 x 1 kg |
£198.81
Pack of 6 |
|
||||
15660750
|
Honeywell Fluka
04273-6X2.5KG
|
6 x 2.5 kg |
£345.51
Pack of 6 |
|
||||
15640750
|
Honeywell Fluka
04273-25KG
|
25 kg |
£225.48
25kg |
|
Sodium phosphate dibasic dodecahydrate, meets analytical spec. of BP, Ph. Eur., E339, 98.5-101% (calc. to the dried substance), Honeywell Fluka™
meets analytical specification of BP, European Pharmacopoeia, E339, 98.5-101% (calc. to the dried substance)
£35.45 - £345.51
Chemical Identifiers
CAS | 10039-32-4 |
---|---|
Molecular Formula | H25Na2O16P |
Molecular Weight (g/mol) | 358.137 |
MDL Number | MFCD00149181 |
InChI Key | DGLRDKLJZLEJCY-UHFFFAOYSA-L |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
PubChem CID | 61456 |
IUPAC Name | disodium;hydrogen phosphate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] |
Specification
Chemical Name or Material | Sodium Phosphate Dibasic Dodecahydrate |
---|---|
Name Note | Meets analytical specification of Ph.Eur.BP |
CAS | 10039-32-4 |
Quantity | 1 kg |
Molecular Formula | H25Na2O16P |
Linear Formula | Na2HPO4 · 12H2O |
MDL Number | MFCD00149181 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
Synonym | disodium phosphate dodecahydrate,disodium hydrogen phosphate dodecahydrate,phosphoric acid, disodium salt, dodecahydrate,di-sodium hydrogen phosphate dodecahydrate,sodium hydrogenphosphate dodecahydrate,unii-e1w4n241fo,sodium phosphate dibasic dodecahydrate,sodium phosphate, dibasic, dodecahydrate,disodium orthophosphate, dodecahydrate,sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
InChI Key | DGLRDKLJZLEJCY-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] |
IUPAC Name | disodium;hydrogen phosphate;dodecahydrate |
Molecular Weight (g/mol) | 358.137 |
PubChem CID | 61456 |
Formula Weight | 358.14g/mol |
Percent Purity | 98.5 to 101% (calc. to the dried substance) |