missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dodecahydrate, meets analytical spec. of BP, Ph. Eur., E339, 98.5-101% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15650750 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
Pack of 6
This item is not returnable. View return policy
This item is not returnable. View return policy

meets analytical specification of BP, European Pharmacopoeia, E339, 98.5-101% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.137
MDL Number MFCD00149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 6 x 1 kg
Linear Formula Na2HPO4 · 12H2O
Formula Weight 358.14
Percent Purity 98.5 to 101% (calc. to the dried substance)
Chemical Name or Material Sodium Phosphate Dibasic Dodecahydrate, Meets analytical specification of Ph.Eur.BP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.