missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BDNF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£261.00 - £5550.00
Specifications
Accession Number | P23560 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 627 |
Molecular Weight (g/mol) | 13-14 kDa, under reducing conditions. |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived BDNF protein His129-Arg247 Accession &Num; P23560 100% sequence homology with Mouse,Rat,Canine,Equine and all other mammalian proteins examined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18727056
|
R&D Systems™
248-BDB-005 |
5 μg |
£261.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18719356
|
R&D Systems™
248-BDB-010 |
10 μg |
£349.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763198
|
R&D Systems™
248-BDB-050 |
50 μg |
£760.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18738806
|
R&D Systems™
248-BDB-250 |
250 μg |
£2140.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728716
|
R&D Systems™
248-BDB-01M |
1 mg |
£5550.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human BDNF Protein is derived from Sf 21 (baculovirus). The Recombinant Human BDNF Protein has been validated for the following applications: Bioactivity.Specifications
P23560 | |
627 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived BDNF protein His129-Arg247 Accession &Num; P23560 100% sequence homology with Mouse,Rat,Canine,Equine and all other mammalian proteins examined. | |
RUO | |
BDNF | |
Human, Mouse, Rat, Canine, Equine | |
Reconstitute 5 μg vials at 100 μg/mL in water. Reconstitute 10 μg or larger vials at 250 μg/mL in water. | |
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining. |
Bioactivity | |
13-14 kDa, under reducing conditions. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Unconjugated | |
Recombinant | |
Lyophilized from a 0.2 μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title