4440.00 GBP valid until 2025-07-30
Use promo code "24078" to get your promotional price.
Use promo code "24078" to get your promotional price.
Accession Number | P23560 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 627 |
Molecular Weight (g/mol) | 13-14 kDa, under reducing conditions. |
Quantity | 1 mg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived BDNF protein His129-Arg247 Accession &Num; P23560 100% sequence homology with Mouse,Rat,Canine,Equine and all other mammalian proteins examined. |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
Regulatory Status | RUO |
Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
Specific Reactivity | BDNF |
Conjugate | Unconjugated |
Species | Human,Mouse,Rat,Canine,Equine |
Species | Human,Mouse,Rat,Canine,Equine |
Species | Human,Mouse,Rat,Canine,Equine |
Species | Human,Mouse,Rat,Canine,Equine |
Species | Human,Mouse,Rat,Canine,Equine |
Recombinant | Recombinant |
Reconstitution | Reconstitute 5 μg vials at 100 μg/mL in water. Reconstitute 10 μg or larger vials at 250 μg/mL in water. |
Buffer | Lyophilized from a 0.2 μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein. |
Purity or Quality Grade | >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining. |