missing translation for 'onlineSavingsMsg'
Learn More

uvex Profabutyl B-05R Chemical Protection Gloves

Constructed of 100% butyl rubber with 0.50 mm bromobutyl coating for chemical protection when working with polar compounds such as esters and ketones.

Brand:  uvex 6094911

42.54 GBP valid until 2025-10-31
Use promo code "24133" to get your promotional price.


 View more versions of this product

Product Code. 17280501

  • £52.80 / 1 pair
Save Big on Hand Protection
Use promo code 24133 at checkout
View Available Promotions
Only null left
Estimated Shipment: 30-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Butyl rubber chemical protection gloves without stockinette
  • Seamlessly coated with 0.50 mm bromobutyl
  • Enhanced grip in damp or wet applications
  • Resistant to polar compounds such as esters, ketones, aldehydes, acids, and alkalis
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Laboratory
Black
Butyl Rubber
11
Type A
Anti-Static Areas, Chemical Handling
Gauntlet
35 cm
Bromobutyl
Yes
Butyl Gloves
2010X
ABIKLNOT
Unisex
Profabutyl B-05R
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.