missing translation for 'onlineSavingsMsg'
                
Learn More
Learn More
Alpha SMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55135-1-AP-20UL
                        
                            
                        
                        
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
This antibody stains the aorta only.
Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predomitly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.Specifications
| Alpha SMA | |
| Polyclonal | |
| Unconjugated | |
| ACTA2 | |
| a SMA, AAT6, ACTA2, ACTA2 Smooth muscle Actin, ACTA2 Smooth muscle Actin Specific, Actin, aortic smooth muscle, Alpha actin 2, Alpha SMA, Alpha SMA Specific, a-SMA, smooth muscle actin, alpha SMA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11475, 59, 81633 | |
| -20°C | |
| Liquid | 
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62736, P62737, P62738 | |
| ACTA2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                             
                            