Invitrogen™ iBlot™ 3 Transfer Stacks, midi, PVDF
iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments.
Additional Details:
Weight:
1.30000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
| Length (Metric) | 13.5 cm |
|---|---|
| Width (Metric) | 8 cm |
| Quantity | 10 Stacks |
| Format | Transfer Stack |
| Material | PVDF |
| Pore Size | 0.2 μm |
| Content And Storage | Store at room temperature. |
| Dimensions (LxW) | 13.5 x 8 cm (Midi) |
| Product Line | iBlot |
| Sufficient For | 10 Blots |