missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 3 Transfer Stacks, midi, PVDF
GREENER_CHOICE

Product Code. p-200000345
Change view
Click to view available options
Quantity:
10 Stacks
3 x 10 Stacks
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17846681 10 Stacks Pack of 10
18495952 3 x 10 Stacks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17846681 Supplier Invitrogen™ Supplier No. IB34001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments.

iBlot 3 Transfer Stacks are used with the iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments. iBlot 3 Transfer Stacks contain copper-fiber electrode sheets, optimized cathode and anode solid buffer gels, and a preconditioned PVDF transfer membrane. They are compatible with common commercially available protein gels, including Bis-Tris, Tris-Glycine, Tris-Acetate, and Tricine chemistries. This midi-size stack (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels.

The iBlot 3 Western Blot Transfer System (iBlot 3 Transfer Stacks plus iBlot3 Transfer Device) has been optimized to provide protein transfers in 3–8 minutes that produce higher quality and more reliable western blot result compared to many other transfer methods.

PVDF membrane

The PVDF membrane (0.2-μm pore size, low fluorescence) in this stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. Proteins bind to the membrane due to hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 13.5 cm
Width (Metric) 8 cm
Quantity 10 Stacks
Format Transfer Stack
Material PVDF
Pore Size 0.2 μm
Content And Storage Store at room temperature.
Dimensions (LxW) 13.5 x 8 cm (Midi)
Product Line iBlot
Sufficient For 10 Blots

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.