Code:
16
Hamilton™ PRP™Series Guard Cartridge Replacement Cartridges
Replacement Cartridges for PRP-1, PRP-3, PRP-X100, PRP-X200, PRP-X300, PRP-X400
Additional Details:
Weight:
0.04200kg
Specification
| pH | 1 to 13 |
|---|---|
| Particle Size | 10 μm |
| Length (Metric) | 25 mm |
| Max. Pressure | 5000 psi |
| Diameter (Metric) Inner | 2.0 mm |
| For Use With (Equipment) | Organic Acid Analysis Column |
| Stationary Phase | C18 |
| Material | 316 Stainless Steel |
| Sorbent | C18 |