Proteintech YPEL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Additional Details:
Additional Details: Weight: 0.00010kg



Product Code. 16849003

Quantity Price
1 £ 349.0 / 150µL
In stock, ready to ship: 2
Add to basket


Description and Specification

Specification

Antigen YPEL5
Applications Immunohistochemistry (Paraffin),Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene YPEL5
Gene Accession No. P62699, P62700
Gene Alias CGI 127, Protein yippee like 5, yippee like 5 (Drosophila), YPEL5
Gene Symbols YPEL5
Host Species Rabbit
Immunogen YPEL5 Fusion Protein Ag2328
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298792, 383295, 51646
Target Species Human,Mouse,Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG

YPEL5 (yippee-like 5) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL5 is expressed at the nucleus and centrosome during interphase; during mitosis, it localizes to the spindle poles, mitotic spindle, and spindle midzone during mitosis. Finally, during cytokinesis, YPEL5 is localized to the midbody. It is associated with the Ran Binding Protein in the Microtubule organizing center (RanBPM) and a related protein RanBP10.