41.21 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Color | Red |
---|---|
Material | Polypropylene |
No. per Case | 100 Cs. |
Neck Finish | 20-410 |
Certifications/Compliance | ISO 9001 |
Product Type | Polypropylene Cap |
For Use With (Equipment) | Plastic dropping bottle |