Waters Corporation XSelect HSS C18 Column
The HSS C18 is a general-purpose C18 bonded phase column that provides superior peak shape for bases, increased retention, and exceptional column lifetimes at low pH.
Additional Details:
Weight:
0.10200kg
Specification
Diameter (Metric) | 4.6 mm |
---|---|
Length (Metric) | 30 mm |
Max. Pressure | 6000 psi (415 Bar) |
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 100 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Endcapped | Yes |
Type | Column |