Learn More
Collagen Type VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17023-1-AP-20UL
This item is not returnable.
View return policy
Description
The extensive family of COL gene products(collagens) iscomposed ofseveral chain types, including fibril-forming interstitialcollagens(typesI, II, III and V) and basement membrane collagens(type IV), each type containing multiple isoforms. Collagens are fibrous, extracellularmatrix proteinswith high tensile strength and are themajorcomponents ofconnective tissue,such astendons and cartilage. Allcollagenscontain a triple helix domain and frequentlyshow lateralself-association in order to formcomplexconnective tissues. Several collagens also play a role in cell adhesion, important formaintaining normal tissue architecture and function.Specifications
Collagen Type VI | |
Polyclonal | |
Unconjugated | |
COL6A1 | |
COL6A1, collagen 6A1, Collagen alpha 1(VI) chain, collagen, type VI, alpha 1, OPLL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12833, 1291, 294337 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12109, Q04857 | |
COL6A1 | |
Collagen Type VI Fusion Protein Ag10288 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |