Learn More
PURUS™ Critical Print Cleanroom Notebooks
Critical Print Notebooks are ideal for writing and data recording in cleanroom environments.
Brand:  PURUS™ PNBCPA62240 WR
 View more versions of this product
Product Code. 15793609
  • £295.00 / Pack of 40
Only null left
Estimated Shipment: 17-09-2025
Add to basket
Description
Description
For writing in cleanroom environments
  • Cleanroom processed and packaged
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanrooms
  • Proprietary Binding Technology without organic fillers
  • Recyclable
  • College Rule
  • Amide and Silicone Free
Specifications
Specifications
Polyethylene
White
148 x 105 mm
Autoclavable
For writing in cleanroom environments
40/Cs.
80 gsm
Yes
Poly
105 mm
Suitable for use in ISO 4 to 6 (Class 10 to Class 1000)
Cleanroom Notebook
A6