Learn More
Branson Ultrasonics™ Ultrasonic Cleaning Bath Accessory, Solid Insert Tray
Solid tray for indirect cleaning method.
Brand: Branson Ultrasonics™ 100-410-172
88.68 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Solid tray for ultrasonic cleaners to prevent parts from touching tank bottom, also easily cleans discarded soil from solid tray
- Also ideal for sample preparation
Specifications
Solid Insert Tray | |
2.7 L | |
7.62 cm | |
Built in handle | |
19.05 cm | |
7.5 x 3.5 in. (19.05 x 8.89 cm) | |
8.89 cm |
3 in. | |
0.75 gal. | |
Ultrasonic Bath, Model 2800 | |
7.5 in. | |
Stainless Steel | |
3.5 in. |