392.73 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Certifications/Compliance | 21 CFR 820 Quality System Regulations,AS/NZS 4179,ASTM D3577,ASTM D7160 |
---|---|
Certifications/Compliance | 21 CFR 820 Quality System Regulations,AS/NZS 4179,ASTM D3577,ASTM D7160 |
Certifications/Compliance | 21 CFR 820 Quality System Regulations,AS/NZS 4179,ASTM D3577,ASTM D7160 |
Certifications/Compliance | 21 CFR 820 Quality System Regulations,AS/NZS 4179,ASTM D3577,ASTM D7160 |
Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009, EN 556, EN ISO 374-1:2016, EN ISO 374-5:2016 |
Chemotherapeutic Rated | No |
Chlorinated | Yes |
Cleanroom Class | Class 100 (ISO 5) |
Accelerator Free | No |
AQL | 1 |
Coating Material | Latex |
Color | Natural |
Disposable | Single-use |
Exterior Finish | Partially Textured |
Finger Thickness (English) | 9.1 mil |
Finger Thickness (Metric) | 0.23 mm |
Hand | Specific |
Latex Free | No |
Material | Latex |
Palm Thickness (English) | 9.1 mil |
Palm Thickness (Metric) | 0.23 mm |
Powdered | Powder Free |
Size | 6 |
Sterility | Sterile |
Thickness (English) | 9.1 mil |
Thickness (Metric) | 0.23 mm |
Glove Cuff Style | Straight |
Grip Design | Smooth |
Manufacturing/QMS Audit Standards | ISO 9001:2015 |
Model | 91-225 |
Series | AccuTech™ 91-225 |
Tensile Strength | 24 MPa |
Length (English) | 11.8 in. |
Length (Metric) | 30 cm |