Learn More
Bel-Art™ SP Scienceware™ Reusable Polypropylene Scoops
Designed for retrieving samples from a larger source. SP Scienceware™ Reusable Polypropylene Scoops are a valuable multi-purpose lab tool.
Brand:  Bel-Art™ F36748-0000
 View more versions of this product
Product Code. 11791485
  • £38.30 / Pack of 12
Only null left
In Stock
Add to basket
Description
Description
Designed for retrieving samples from a larger source. SP Scienceware™ Reusable Polypropylene Scoops are a valuable multi-purpose lab tool.
  • Rigid form scoop
  • Flat bottom keeps scoops upright when filled or empty
  • Can be used as weighing boat
  • Manufactured from polypropylene
  • Steam autoclavable at 121°C (250°F)
  • Complies with FDA regulations for food contact
  • Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes
  • Stated volumes represent typical heaping samples
Specifications
Specifications
Scoop
Autoclavable
60 mL
One-piece
Smooth
Designed to remain upright when filled or empty allowing them to be used as weighing boats or to be placed on a flat surface for ease sample collection and dispensing
1.12 in.
8.06 in.
20.5 cm
1.62 in.
Polypropylene
2.02 oz.
Reusable
Translucent
FDA
20.5 x 4.1 x 2.9 cm
2.9 cm
5 in.
12.7 cm
4.1 cm