Greiner Bio-One Disposable Inoculation Loops and Needles
Manufactured from polystyrene for strength and flexibility. Greiner Bio-One™ Disposable Inoculation Loops provide easier collection and inocluation,
Additional Details:
Additional Details: Weight: 6.10000kg
Product Code. 10792142
Quantity Price
1 £ 138.93 / Pack of 2000
Estimated Shipment
02-09-2025
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Disposable Single-use
Length (Metric) 200 mm
Material Polystyrene
Sterility Sterile
Color Blue
Product Type Inoculating Needle
Packaging Type Bulk
  • Needles are ideal for picking up single colonies; Loops are ideal for picking up designated volume of culture
  • Packaged individually or bulk
  • Hydrophobic
  • Temperature range: –20 to +60°C
  • Sterilized by irradiation
  • For single use only
Alternative Products