Code:
94
DuPont™ Tyvek 500 Industry Coveralls
Provides an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 500 Industry Coveralls with Collar are composed of flash spun high density polyethylene.
Additional Details:
Weight:
0.13500kg
Specification
Product Type | Collared Coveralls |
---|---|
Size | Small |
Material | HDPE |
Color | White |
Waist Style | Elastic (Stitched-in) |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Collared |
Closure Type | Zipper with Storm Flap |
Seam Type | Stitched |
Protection Class | Category III, Type 5 and 6 |
Collar Type | Standard fold |
Certifications/Compliance | EN 1073-2, EN 1149-5 |
Size (Metric) Chest | 84 to 92 cm |
For Use With (Application) | For industrial applications |
Height (Metric) | 162 to 170 cm |
Quantity | 100/Pk. |