Learn More
R&D Systems™ Recombinant Human IL-6 Protein
A New rh IL-6 is Available! The new protein has ∼2 fold better activity and is HEK293 expressed!
Brand: R&D Systems™ 206-IL-010

Description
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.2-0.8 ng/mL. The specific activity of Recombinant Human IL-6 is approximately 1.1 x 105 IU/μg, which is calibrated against human IL-6 WHO International Standard (NIBSC code: 89/548). Specific activity is for reference purposes only and is not routinely tested.Specifications
Q75MH2 | |
3569 | |
IL-6 | |
E. coli-derived human IL-6 protein Pro29-Met212 | |
<0.10 EU / 1 µg of the protein by the LAL method. | |
IL6 | |
Yes |
Lyophilized from a 0.2 µm filtered solution in PBS and NaCl with BSA as a carrier protein. | |
M.W. theroretical: 20.9 kDa | |
10 µg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
Unconjugated | |
Reconstitute at 100-200 µg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.