Memmert™ Software Conforming to FDA AtmoCONTROL
Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Additional Details:
Additional Details: Weight: 0.01100kg
Product Code. 17189834
Quantity Price
1 £ 2240.0 / Each
Estimated Shipment
17-10-2025
Add to basket
Description and Specification
Specification
For Use With (Application) Only for Units with TwinDISPLAY
For Use With (Equipment) HPP410eco
  • Base licence for reading the data logging of one unit (only for units with TwinDISPLAY)
  • Respective IQ/OQ documents available in German and English language (without surcharge)