Learn More
Memmert™ Software Conforming to FDA AtmoCONTROL
Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Brand: Memmert™ IPP1060ecoFDAQ1
This item is not returnable.
View return policy
Description
- Base licence for reading the data logging of one unit (only for units with TwinDISPLAY)
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Only for Units with TwinDISPLAY | |
IPP1060eco |