uvex Rubiflex S NB60S Chemical Protection Gloves
Long, NBR-coated chemical protection gloves feature reinforced cotton interlock supporting material for applications that require protection against chemicals and mechanical hazards.
Additional Details:
Weight:
0.30200kg
Specification
Applications | Laboratory |
---|---|
Coating Material | NBR |
Color | Green |
Latex Free | Yes |
Material | Nitrile |
Product Type | Chemical-resistant Gloves |
Size | 11 |
EN388 Standard | 2121X |
EN ISO 374-1 | Type B |
EN ISO 374-1 CHEMICAL LETTER CODES | JKOPT |
For Use With (Application) | Chemical Handling, Municipal Cleaning, Sandblasting, and Sewer Construction |
Gender | Unisex |
Glove Cuff Style | Gauntlet |
Liner Material | Cotton |
Series | Rubiflex S NB60S |