missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TNF-alpha Protein
Click to view available options
Quantité:
5 μg
Conditionnement:
5µg
Description
Measured in a cytotoxicity assay using L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D. Matthews, N. and M.L. Neale (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 221. The ED50 for this effect is 25-100 pg/mL. The specific activity of Recombinant Human TNF-alpha is approximately 7.6 x 104 IU/μg, which is calibrated against human TNF-alpha WHO International Standard (NIBSC code: 88/786). Specific activity is for reference purposes only and is not routinely tested.

Spécification
Spécification
Formule | Lyophilized from a 0.2μm filtered solution in PBS. |
Identification génétique (Entrez) | 7124 |
Poids moléculaire | 17.5 kDa |
Nom | TNF-alpha |
Quantité | 5 μg |
Source | E. coli-derived human TNF-alpha protein Val77-Leu233, with and without an N-terminal Met |
Concentration en endotoxines | Measured in a cytotoxicity assay using L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D. Matthews, N. and M.L. Neale (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 221. The ED50 for this effect is 25-100 pg/mL. |
Alias de gène | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
Conjugué | Unconjugated |
Recombinant | Recombinant |
Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu