missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta 1 Protein
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Formule:
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Conditionnement:
10µg
1mg
2µg
Description
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.04-0.2 ng/mL.

Spécification
Spécification
Formule | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
Identification génétique (Entrez) | 7040 |
Poids moléculaire | 12.8 kDa (monomer) |
Nom | TGF-beta 1 |
Quantité | 10 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Ala279-Ser390 |
Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Forme structurelle | Disulfide-linked homodimer |
Alias de gène | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
Conjugué | Unconjugated |
Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu