missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IGF-I/IGF-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£236.00 - £620.00
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
---|---|
Gene ID (Entrez) | 3479 |
Molecular Weight (g/mol) | 7.6 kDa |
Name | IGF-I/IGF-1 |
Source | E. coli-derived human IGF-I/IGF-1 protein Gly49-Ala118 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18196850
|
R&D Systems™
291-G1-200 |
200 μg |
£236.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18117990
|
R&D Systems™
291-G1-01M |
1 mg |
£620.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 0.3-1.5 ng/mL.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
7.6 kDa | |
E. coli-derived human IGF-I/IGF-1 protein Gly49-Ala118 | |
Unconjugated | |
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
3479 | |
IGF-I/IGF-1 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title