208.80 GBP valid until 2025-07-30
Use promo code "24078" to get your promotional price.
Use promo code "24078" to get your promotional price.
Accession Number | Q75MH2 |
---|---|
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein. |
Gene ID (Entrez) | 3569 |
Molecular Weight (g/mol) | M.W. theroretical: 20.9 kDa |
Name | IL-6 |
Quantity | 10 μg |
Source | E. coli-derived human IL-6 protein Pro29-Met212 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Gene Alias | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
Gene Symbol | IL6 |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Reconstitution | Reconstitute at 100-200 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.) |
Purity or Quality Grade | >97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Protein | IL-6 |