Learn More
3M™ 4530 Protective Polypropylene Coveralls with Hood
Made from super-breathable 54 gsm SMS non-woven polypropylene, these coveralls offer excellent barrier protection against dry particles (inc. Cat. III hazardous dusts) and certain liquid-chemical splash hazards. 4530 Protective Coveralls with Hood includes 3-panel hood, 2-way zip and storm flap, and elasticated waist and ankles.
Brand: 3M™ 7000109014
128.71 GBP valid until 2025-09-30
Use promo code "24380" to get your promotional price.
Description
- Product Type: Protective Coverall
- Material: SMS Non-Woven Polypropylene
- Hood: 3-Panel
- Closure: 2-Way Zip and Storm Flap
- Seams: Serged
- Waist: Elasticated
- Ankles: Elasticated
- Cuff: Knitted
- Weight: 54 gsm
- Protection: Nuclear Particulate Resistance (EN 1073-2), Type 5 Dust (EN ISO 13982-1), Type 6 Liquid Splash (EN 13034), Flame-Spread Resistant (ISO 14116 Index 1), and Antistatic (EN 1149-5)

Specifications
Protective Coveralls | |
Unisex | |
Blue | |
Elastic | |
Knit | |
3-panel | |
Serged | |
Category III | |
Nuclear Particulate Resistance (EN 1073-2), Type 5 Dust (EN ISO 13982-1), Type 6 Liquid Splash (EN 13034), Flame-Spread Resistant (ISO 14116 Index 1), and Antistatic (EN 1149) | |
45.3 to 48.8 in. | |
73.2 to 76.4 in. | |
54 gsm |
2X-Large | |
SMS Polypropylene | |
Single-use | |
Elastic | |
Hooded | |
2-way Zipper with Storm Flap | |
Non-sterile | |
Flame, Chemicals, Liquid, Anti-static | |
None | |
115 to 124 cm | |
186 to 194 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.