missing translation for 'onlineSavingsMsg'
Learn More

Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™

Product Code. 17368573 Shop All MP Biomedicals Products
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Pen-Strep (10,000 IU/mL, 10 mg/mL) is a double antibiotic solution containing 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.

  • The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
  • This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Concentration 10,000 IU/mL Penicillin, 10 mg/mL Streptomycin
Quantity 20 mL

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.