missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tricine Mini Protein Gels, 10%, 1.0 mm

Product Code. p-7237826
Click to view available options
Wells:
10-well
12-well
Unit Size:
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels.

Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. In this system tricine replaces glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.

Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow (Schaegger and von Jagow, 1987) specifically for resolving peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.

Features of Novex Tricine protein gels:
• Increased resolution of proteins with molecular weights as low as 2 kDa
• Improved compatibility with direct sequencing of proteins after transferring to PVDF
• Minimized protein modification due to the lower pH of the tricine buffering system

Formulation
Invitrogen Tricine gels are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, and highly purified water. Our Tricine gels have a 4% stacking gel and do not contain SDS. The Tricine system requires SDS in sample and running buffers for best results.

Choose the right Tricine gel for your protein separation
Invitrogen Tricine gels come in three polyacrylamide concentrations of 10%, 16%, and a gradient of 10–20%. Select from our many well formats, including 10-, 12-, and 15-well. Tricine gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, we recommend Tricine SDS Sample Buffer (LC1676) and optimal separation use Tricine SDS Running Buffer (LC1675).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) if performing a traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 10%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tricine
Recommended Applications Denaturing
Sample Loading Volume Up to 25 μL
Separation Range 6 to 220 kDa
Separation Type Denaturing
Wells 10-well
Mode of Separation Molecular Weight
Product Line Novex™
Quantity 10 Gels/Box
Shipping Condition Wet Ice
Storage Requirements Store at 2°C to 8°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.