Micronova™ NovaClean™ Lab and Glass Cleaner

Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.

£294.0 - £473.0

Specification

Product Type Filtered Low Ionic Surfactant (0.1 μ)
Color-coded No
pH 7.1
Product Line NovaClean
Scent Unscented
View More Specs


Specification

Product Type Filtered Low Ionic Surfactant (0.1 μ)
Color-coded No
pH 7.1
Product Line NovaClean
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Boiling Point 99°C (210.2°F)
Care Instructions Avoid breathing vapor or mist and provide adequete ventilation when using.
Certifications/Compliance ISO 9001 Certified Facility; Certificate of Analysis
Color Clear
Content And Storage Store at ambient temperature. Avoid direct sunlight. Store in original container and keep tightly closed and seaqled until ready for use.
Density 1.025 g/mL at 25°C
Disposable Reusable
Filtered Filtered
Flammable No
For Use With (Application) Ready to use filtered cleanroom detergent, no mobile ions, free-rinsing for cleaning glass, stainless steel, acrylic and painted surfaces.
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Melting Point 0°C (32°F)
Resistant To The product is chemically stable. Under normal conditions of storage and use, hazardous reactions will not occur.
Vapor Pressure 2.3 kPa (17.1 mmHg)

Lab and glass cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.

  • Ready to use, cleanroom surfactant
  • Filtered to 0.1 microns
  • Low mobile ions
  • Low non-volatile residue levels
  • Can be applied directly to a range of surfaces
  • Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
  • Ideal for conductive flooring
  • Available in a 1 quart spray bottle and a 1 gallon refill bottle
  • Biodegradable
  • Free-rinsing

  Product Code Brand Volume (English) Price ??product.searchResults.label.quantityAvail??
Quantity
11962475
Micronova™
NC2-G
1 gal. £473.0
Pack of 4
Add to basket
** 11966855
Micronova™
NC2-Q
1 qt. n/a  
** No longer available. See item for replacement.