Invitrogen™ Human NGF beta ELISA Kit
ELISA
Additional Details:
Weight:
0.33820kg
Specification
Accession Number | P01138 |
---|---|
Assay Range | 6.86 to 5000 pg/mL |
Assay Sensitivity | 14 pg/mL |
Calibration Range | 14 pg/mL to 5000 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | HRP |
Product Type | ELISA |
Protein Family | Growth Factors & Receptors |
Sample Type | Plasma,Serum,Supernatant |
Sample Type | Plasma,Serum,Supernatant |
Sample Type | Plasma,Serum,Supernatant |
Specificity | This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, MMP-1, - 2, -3, -10, PARC, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF |
Sufficient For | 5 x 96 Assays |
Target Species | Human |
For Use With (Application) | ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
For Use With (Equipment) | Colorimetric Microplate Reader |
Gene ID (Entrez) | 4803 |
Gene Symbol | Beta-NGF,HSAN5B,NGF |
Interassay CV | <12% |
Intraassay CV | <10 |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Kit Contents | Pre-coated 96 well plate,Standard,Assay Diluent concentrate,Biotinylated Detection Antibody,SAV-HRP,Wash Buffer,Chromogen,Stop Solution,Adhesive Plate Covers |
Label or Dye | HRP |
Quantity | 5 x 96 Tests |
Regulatory Status | RUO |
Research Discipline | Neurobiology |
Sample Volume | Plasma 50 μL, Serum 50 μL, Supernatant 100 μL |
Storage Requirements | 2°C to 8°C |
Synonym | Beta-NGF, HSAN5B, NGF |
Target | NGF |
Test Time | 1 hrs 20 mins |
Total Assay Time | 4 hrs 45 mins |