missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22280-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can specifically recognize endogenous MYH7, not cross-reacting with other MHCs.
Muscle myosin is a hexameric protein containing 2 heavy chain subunits, 2 alkali light chain subunits, and 2 regulatory light chain subunits. This gene encodes the beta (or slow) heavy chain subunit of cardiac myosin. It is expressed predomitly in normal human ventricle. It is also expressed in skeletal muscle tissues rich in slow-twitch type I muscle fibers. Changes in the relative abundance of this protein and the alpha (or fast) heavy subunit of cardiac myosin correlate with the contractile velocity of cardiac muscle. Its expression is also altered during thyroid hormone depletion and hemodynamic overloading. Mutations in this gene are associated with familial hypertrophic cardiomyopathy, myosin storage myopathy, dilated cardiomyopathy, and Laing early-onset distal myopathy.Specifications
| MYH7 | |
| Polyclonal | |
| Unconjugated | |
| MYH7 | |
| beta MHC, CMD1S, CMH1, MPD1, MYH7, MyHC beta, MyHC slow, MYHCB, Myosin 7, Myosin heavy chain 7, SPMD, SPMM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140781, 4625 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12883, Q91Z83 | |
| MYH7 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction