missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Macro Pipetters
GREENER_CHOICE

Product Code. 11915842
Click to view available options
Volume (Metric):
1 mL
2 mL
3 mL
4 mL
5 mL
6 mL
7 mL
8 mL
9 mL
10 mL
Accuracy:
±0.6%
±0.75%
Color:
Gray
Green
Red
White
Yellow
Unit Size:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

For large volume handling tasks such as reconstituting reagents, controls, or dilutions

  • Factory calibrated for accurate volume delivery
  • Can be easily adjusted to achieve accurate performance on dense or viscous liquids

Specifications

Accuracy ±0.6%
Autoclavable Not Autoclavable
Color Red
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum, Stainless Steel
Model F1
Number of Channels 1
Sterility Non-sterile
Type Macropipette
Volume (Metric) 8 mL
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Lifetime
Length (English) 11 in.
Width (English) 2.5 in.
Height (English) 1.5 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.