VistaLab™ Technologies MLA™ Brand Macro Pipetters

For large volume handling tasks such as reconstituting reagents, controls, or dilutions

Additional Details:
Additional Details: Weight: 0.50000kg



Product Code. 11905822

Quantity Price
1 £ 335.0 / Each
Estimated Shipment
20-11-2025
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Accuracy ±0.75%
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum, Stainless Steel
Model F1
Number of Channels 1
Sterility Non-sterile
Type Macropipette
Volume (Metric) 1 mL
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Lifetime
Length (English) 11 in.
Width (English) 2.5 in.
Height (English) 1.5 in.

  • Factory calibrated for accurate volume delivery
  • Can be easily adjusted to achieve accurate performance on dense or viscous liquids

Alternative Products