Learn More
Fisherbrand™ Mixed Heating (Steam or Electrical)
Mixed heating from a steam source for water heating and an electrically powered source for drying.
£2585.00 - £2970.00
Specifications
For Use With (Application) Steam or Electrical Heating
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17117794
View Documents
Fisherbrand™
2908157
Blue Line 5 / IQ5
£2585.00
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 16-10-2025
Add to basket
17117834
View Documents
Fisherbrand™
2918157
Blue Line 6 / IQ6
£2970.00
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 16-10-2025
Add to basket
Description
Description
  • The energy for water heating comes from steam source
  • The energy for drying is electrically powered
  • If the steam fails, the machine continues to run on electrical power
  • The machine can be programmed to run on steam in certain periods and electricity at others
Specifications
Specifications
Steam or Electrical Heating