missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals LabCare Incubator Water Disinfectant
Maintenance of water and prevents cross-contamination in incubators.
£91.00 - £401.00
Specifications
Product Type | Disinfectant |
---|---|
Color-coded | No |
Form | Liquid |
Scent | Unscented |
Sterility | Non-sterile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
30281206
|
MP Biomedicals
093051249 |
100 mL |
£91.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30281210
|
MP Biomedicals
093051254 |
500 mL |
£401.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- MP Biomedicals LabCare Incubator Water Disinfectant Solution offers a comprehensive solution for maintaining water cleanliness and preventing cross-contamination within incubators
- This high-efficiency, broad-spectrum antimicrobial solution effectively inhibits and eliminates microorganisms present in incubator water
- With its stable formulation and low toxicity, it ensures a clean and sterile environment conducive to optimal incubation conditions
Specifications
Disinfectant | |
Liquid | |
Non-sterile |
No | |
Unscented |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- LabCare Incubator Water Disinfectant Solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 1,2-benzisothiazol-3(2H)-one, benzyldodecyldimethylammoniumbromide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title