Axygen™ HybridRack™ Pipet Tip, Extended Length

The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.

Overview
77.67 GBP valid until 2025-08-22
Use promo code "24148" to get your promotional price.



Additional Details:
Additional Details: Weight: 1.20000kg



Product Code. 30090450

Quantity Price
1 £ 120.96000000000001 / Pack of 960
In stock, ready to ship: 30
Add to basket


Description and Specification

Specification

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Blue
Autoclavable Not Autoclavable
Compatibility Universal
Format Rack
Tip Style Extended Length
Product Type Pipet Tip
Color Code Blue
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene

The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.

  • The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
  • The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
  • The paperboard base can be easily recycled through most local curbside recycling programs.
  • All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
  • The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
  • The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.

Special Offers