Proteintech HNRNPA1 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Additional Details:
Additional Details: Weight: 0.00010kg



Product Code. 16868073

Quantity Price
1 £ 135.0 / 20µL
In stock, ready to ship: 2
Add to basket


Description and Specification

Specification

Antigen HNRNPA1
Applications Immunohistochemistry (Paraffin),Immunofluorescence,Western Blot,Immunocytochemistry
Applications Immunohistochemistry (Paraffin),Immunofluorescence,Western Blot,Immunocytochemistry
Applications Immunohistochemistry (Paraffin),Immunofluorescence,Western Blot,Immunocytochemistry
Applications Immunohistochemistry (Paraffin),Immunofluorescence,Western Blot,Immunocytochemistry
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HNRNPA1
Gene Accession No. P09651, P49312
Gene Alias Helix destabilizing protein, hnRNP A1, hnRNP core protein A1, HNRNPA1, HNRPA1
Gene Symbols HNRNPA1
Host Species Rabbit
Immunogen HNRNPA1 Fusion Protein Ag1656
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15382, 3178, 768103
Target Species Mouse,Human,Pig
Target Species Mouse,Human,Pig
Target Species Mouse,Human,Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA-binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has four repeats of quasi-RNA recognition motif (RRM) domains that bind RNAs. This protein binds to the intronic polypyrimidine tracts that requires pre-mRNA splicing and acts via the protein degradation ubiquitin-proteasome pathway. It may also promote the binding of U2 snRNP to pre-mRNAs. This protein is localized in the nucleoplasm and it is also detected in the perinucleolar structure. Alternatively spliced transcript variants encoding different isoforms have been described.