Fisherbrand™ High Speed Module
High speed module consist of 2 tanks on top of the machine.
Overview
Additional Details:
Additional Details: Weight: 8.00000kg
Product Code. 17197804
Quantity Price
1 £ 5485.0 / Each
Estimated Shipment
13-08-2025
Add to basket
Description and Specification
Specification
Product Type High Speed Module
For Use With (Equipment) Blue Line 5 / IQ5
Includes 2 Tanks
  • Tanks electrically heated
  • Water is preheated before going into the washing chamber, allowing for shorter cycle times