Glycerol, ≥99.5%, Molecular biology reagent grade, MP Biomedicals™

Glycerol is a simple polyol compound which is a viscous liquid that is widely used in pharmaceutical formulations. It has three hydroxyl groups that are responsible for its solubility in water and its hygroscopic nature

£77.2 - £164.0

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs


Specification

Chemical Name or Material Glycerol
CAS 56-81-5
Density 1.2613g/cm3
Assay Percent Range ≥99.5%
Molecular Formula C3H8O3
MDL Number MFCD00004722
Quantity 500 mL
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
Synonym glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
PubChem CID 753
ChEBI CHEBI:17754
Formula Weight 92.09
Grade Molecular Biology Reagent
Physical Form Liquid

Product Code Brand Quantity Price ??product.searchResults.label.quantityAvail??
Quantity
11486441
MP Biomedicals
0219399690
500 mL £77.2
500mL
Add to basket
11496441
MP Biomedicals
0219399691
1 L £164.0
1 L
Add to basket