missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Silica
Shop All Buchi Products
Click to view available options
Includes:
10 Pc.
12 Pc.
18 Pc.
20 Pc.
5 Pc.
6 Pc.
Bed Weight (Metric):
4 g
12 g
25 g
40 g
80 g
220 g
330 g
Unit Size:
Pack of 10
Pack of 12
Pack of 18
Pack of 20
Pack of 5
Pack of 6
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Specifications
Material | Polypropylene |
Type | Silica |
Includes | 5 Pc. |
Resolution | 40 to 63 μm |
Bed Weight (Metric) | 330 g |
Column Type | Silica |
For Use With (Application) | Basic Applications, Polar Compounds |
For Use With (Equipment) | Any Flash System |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction