| Product Code | Brand | Quantity | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
|---|---|---|---|---|---|---|---|---|
|
11465461
|
MP Biomedicals
0215804080
|
100 g |
£25.2
100g |
|
||||
|
17897486
|
MP Biomedicals
0215804090
|
500 g |
£61.8
500g |
|
Ferric ammonium citrate, MP Biomedicals™
£25.2 - £61.8
Chemical Identifiers
| CAS | 1185-57-5 |
|---|---|
| Molecular Formula | C6H11FeNO7+3 |
| Molecular Weight (g/mol) | 264.999 |
| InChI Key | FRHBOQMZUOWXQL-UHFFFAOYSA-N |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| PubChem CID | 118984355 |
| IUPAC Name | azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) |
| SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] |
Specification
| Chemical Name or Material | Ferric ammonium citrate |
|---|---|
| CAS | 1185-57-5 |
| Color | Rust to Brown |
| Density | 7.87g/cm3 at 25°C |
| Molecular Formula | C6H11FeNO7+3 |
| Quantity | 100 g |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| Synonym | ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz |
| InChI Key | FRHBOQMZUOWXQL-UHFFFAOYSA-N |
| SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] |
| IUPAC Name | azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) |
| Molecular Weight (g/mol) | 264.999 |
| PubChem CID | 118984355 |
| Formula Weight | 265 |
| Physical Form | Powder |
Product Identifier
- Ferric Ammonium Citrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
Precautionary Statement
- P101- If medical advice is needed, have product container or label at hand.
- P102- Keep out of reach of children.
- P103- Read label before use.
- P264- Wash thoroughly after handling.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321- Specific treatment (see on this label).
- P332+P313- If skin irritation occurs: Get medical advice/attention.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : ammonium iron(III) citrate