Fisherbrand™ Conductivity Sensor
The machine can be equipped with conductivity control.
Additional Details:
Additional Details: Weight: 0.08000kg
Product Code. 17107784
Quantity Price
1 £ 2000.0 / Each
Estimated Shipment
24-12-2025
Add to basket
Description and Specification
Specification
Product Type Conductivity Sensor
  • Conductivity for a final rinse can be set and the number of attempts the machine must try to reach the set value
  • Conductivity control is easily set in the programming of the rinse phase