Fisherbrand™ Condenser Kit for Condenser Recycle
Standard condenser that collects water in a tank rather than letting the water out into the drain.
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 17197844
Quantity Price
1 £ 215.0 / Each
Estimated Shipment
17-09-2025
Add to basket
Description and Specification
Specification
Product Type Condenser Kit
For Use With (Equipment) Condenser Recycle
  • Collected water can then be used for washing in the next cycle
  • Tank includes a heating element, which keeps the temperature above 60°C and ensures that there is no possibility to develop bacteria in the tank