Learn More
C4BPA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11819-1-AP-150UL
 View more versions of this product
Product Code. 16869143
  • £349.00 / 150µL
Description
C4BP is composed of two polypeptides (alpha and beta chains), which have molecular weights of 70 and 45 kDa, respectively. This polyclonal antibody is raised against the C4BP alpha chain (C4BPA).
This gene encodes a member of a superfamily of proteins composed predomitly of tandemly arrayed short consensus repeats of approximately 60 amino acids. Along with a single, unique beta-chain, seven identical alpha-chains encoded by this gene assemble into the predomit isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Two pseudogenes of this gene are also found in the cluster.
Specifications
Specifications
C4BPA
Polyclonal
Unconjugated
C4BPA
C4BP, C4BPA, Proline rich protein, PRP
Rabbit
Antigen Affinity Chromatography
RUO
722
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04003
C4BPA
C4BPA Fusion Protein Ag2387
150 μL