missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Biotin
GREENER_CHOICE

Product Code. p-7230577 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
2 g
5 g
Unit Size:
2g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12853206 2 g 2g
12863206 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12853206 Supplier IBA Lifesciences Supplier No. 21016002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotin powder for preparation of Strep-Tactin™XT elution buffer

  • Biotin is a water-soluble B-complex vitamin, also known as vitamin B7 or vitamin H. It binds to the biotin-binding pockets of streptavidin, Strep-Tactin™, and Strep-Tactin™XT with different affinities.
  • It allows the efficient elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins from Strep-Tactin™XT, while maintaining the reusability of the resin.
  • It can be used for preparation of Strep-Tactin™XT elution buffer, Buffer BXT.
  • In addition, biotin can be used for elution of biotinylated proteins from Strep-Tactin™, where it binds irreversibly to the resin - Strep-Tactin™ cannot be regenerated in this kind of application.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-{2-oxo-hexahydro-1 H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Color White
Quantity 2 g

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.