missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Media Tryptic Soy Broth, Select APS
Click to view available options
:
10kg
Description
- Select Alternative Protein Source (APS)
- Obtained from animal-free components

Specifications
Specifications
Form | Liquid |
Product Type | Microbiology Media |
Packaging Type | Pail |
Quantity | 10,000 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction