Azlon Azlon™ Square Polycarbonate Bottles

Ideal for preparation and storage of biological media, reagents and aqueous solutions

Overview
142.50 GBP valid until 2025-12-31
Use promo code "24373" to get your promotional price.



Code: NEW

Additional Details:
Additional Details: Weight: 2.28000kg



Product Code. 15344941

Quantity Price
1 £ 150.0 / Pack of 12
Estimated Shipment
06-11-2025
Add to basket


Description and Specification

Specification

Capacity (Metric) 500 mL
Diameter (Metric) Inner Neck 28 mm
Material Polycarbonate
Neck ID (Metric) 28mm
Quantity 12/Pk.
Shape Square
Height (Metric) 172 mm
Product Type Square Bottle

  • Highly transparent polycarbonate media bottles with linerless polypropylene cap
  • Suitable for repeated sterilisation by autoclaving at 121°C
  • Leakproof
  • Practical square design for economic use of space
  • Heavy duty and durable in use

Documentation