missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Anodized Aluminum Blocks
Useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial settings. Fisherbrand™ Anodized Aluminum Blocks are for use with Fisherbrand Digital Block Heaters.
Brand: Fisherbrand™ D1125
95.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Excellent heat transfer
- Anodized to resist thermal shock
- Available to fit a range of test tube counts and sizes
Specifications
Holds 6 x 25mm Tubes | |
6 x 25 mm Tubes | |
Block |
Test Tubes | |
Anodized Aluminum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction